Cesium iodide ≥99.999% (Cs basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Cesiumiodide
93-5544-50G
93-5544-10G
100203-516EA
300.06
USD
100203-516
100204-364
Cesium iodide ≥99.999% (Cs basis), PURATREM
Cesium iodide
Formula:
CsI MW: 259.81 g/mol Boiling Pt: 1280 °C (1013 hPa) Melting Pt: 621 °C Density: 3.21 g/cm³ (20 °C) |
MDL Number:
MFCD00010962 CAS Number: 7789-17-5 EINECS: 232-145-2 Merck Index: 12,02056 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...