Cesium iodide ≥99.9% (Cs basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Cesiumiodide

93-5543-25G 93-5543-500G 93-5543-100G
100196-762EA 101.91 USD
100196-762 100199-286 100202-810
Cesium iodide ≥99.9% (Cs basis)
Cesium iodide
Formula: CsI
MW: 259.81 g/mol
Boiling Pt: 1280 °C (1013 hPa)
Melting Pt: 621 °C
Density: 3.21 g/cm³ (20 °C)
MDL Number: MFCD00010962
CAS Number: 7789-17-5
EINECS: 232-145-2
Merck Index: 12,02056

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR