Cesium chloride ≥99.9% (Cs basis)

Supplier: STREM CHEMICALS INC MS

93-5535-10G 93-5535-50G
100205-654EA 42.46 USD
100205-654 100208-192
Cesium chloride ≥99.9% (Cs basis)
Cesium chloride
Formula: CsCl
Boiling Pt: 1290 °C
Melting Pt: 646 °C
Density: 3.988
Storage Temperature: Ambient
MDL Number: MFCD00010955
CAS Number: 7647-17-8
EINECS: 231-600-2
Merck Index: 13,02021

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR