Cesium iodide ≥99% (Cs basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Cesiumiodide
93-5524-500G
93-5524-25G
93-5524-100G
100210-890EA
794.03
USD
100210-890
100205-650
100210-480
Cesium iodide ≥99% (Cs basis)
Cesium iodide
Formula:
CsI MW: 259.81 g/mol Boiling Pt: 1280 °C Melting Pt: 621 °C Density: 3.21 g/cm³ (20 °C) |
MDL Number:
MFCD00010962 CAS Number: 7789-17-5 EINECS: 232-145-2 Merck Index: 12,02056 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...