Cesium hydroxide 50% in aqueous solution ≥99.9% (Cs basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-5520-25G
93-5520-100G
100202-196EA
77.85
USD
100202-196
100199-622
Cesium hydroxide 50% in aqueous solution ≥99.9% (Cs basis)
Cesium hydroxide
Formula:
CsOH MW: 149.91 g/mol |
MDL Number:
MFCD00010964 CAS Number: 21351-79-1 EINECS: 244-344-1 UN: 2681 ADR: 8,II Merck Index: 13,02022 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...