Cesium fluoride ≥99% (Cs basis)

Supplier: STREM CHEMICALS INC MS
Danger

93-5519-25G 93-5519-100G
100196-758EA 55.2 USD
100196-758 100209-152
Cesium fluoride ≥99% (Cs basis)
Cesium fluoride
Formula: CsF
MW: 151.9 g/mol
Boiling Pt: 1251 °C
Melting Pt: 682 °C
Density: 4.627 g/cm³ (20 °C)
MDL Number: MFCD00010960
CAS Number: 13400-13-0
EINECS: 236-487-3
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR