Cesium fluoride ≥99.9% (Cs basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-5518-10G
93-5518-50G
100210-474EA
63.69
USD
100210-474
100207-108
Cesium fluoride ≥99.9% (Cs basis)
Cesium fluoride
Formula:
CsF MW: 151.9 g/mol Boiling Pt: 1251 °C (1013 hPa) Melting Pt: 680 °C Density: 4.627 g/cm³ (20 °C) |
MDL Number:
MFCD00010960 CAS Number: 13400-13-0 EINECS: 236-487-3 UN: 3288 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...