Cesium bromide ≥99% (Cs basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Cesiumbromide

93-5513-25G 93-5513-100G
100207-106EA 82.09 USD
100207-106 100210-472
Cesium bromide ≥99% (Cs basis)
Cesium bromide
Formula: BrCs
MW: 212.81 g/mol
Boiling Pt: 1300 °C
Melting Pt: 636 °C
Density: 4.46 g/cm³ (20 °C)
MDL Number: MFCD00010954
CAS Number: 7787-69-1
EINECS: 232-130-0
Merck Index: 13,02019

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR