Cesium acetate ≥99.9% (Cs basis)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Acetic acid cesium salt
93-5510-10G
93-5510-50G
100196-754EA
76.43
USD
100196-754
100196-756
Cesium acetate ≥99.9% (Cs basis)
Cesium acetate
Formula:
H₃CCOOCs MW: 191.95 g/mol Melting Pt: 194 °C |
MDL Number:
MFCD00013056 CAS Number: 3396-11-0 EINECS: 222-248-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...