Tellurium ≥99.99%, broken ingot
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-5224-25G
93-5224-100G
100205-644EA
99.08
USD
100205-644
100210-468
Tellurium ≥99.99%, broken ingot
Tellurium
Formula:
Te Boiling Pt: 989.8 °C Melting Pt: 450 °C Density: 8.83 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134062 CAS Number: 13494-80-9 EINECS: 236-813-4 UN: 3288 ADR: 6.1,III Merck Index: 12,09273 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...