Tellurium tetrachloride ≥99.9% (Te basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tellurium(IV) chloride

93-5205-50G 93-5205-10G
100196-728EA 233.54 USD
100196-728 100197-888
Tellurium tetrachloride ≥99.9% (Te basis)
Tellurium tetrachloride
Formula: Cl₄Te
MW: 269.41 g/mol
Boiling Pt: 380
Melting Pt: 224
Density: 3.26
Storage Temperature: Ambient
MDL Number: MFCD00011262
CAS Number: 10026-07-0
EINECS: 233-055-6
UN: 1759
ADR: 8 II
Merck Index: 12,09279

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR