Antimony ≥99.9999%, shot
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-5145-5G
93-5145-25G
100209-146EA
93.42
USD
100209-146
100202-562
Antimony ≥99.9999%, shot
Antimony
Formula:
Sb Boiling Pt: 1637 °C (1013 hPa) Melting Pt: 630 °C Density: 6.684 g/cm³ (20 °C) |
MDL Number:
MFCD00134030 CAS Number: 7440-36-0 EINECS: 231-146-5 UN: 2871 ADR: 6.1,III Merck Index: 12,00733 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...