Antimony ≥99.5%, powder

Supplier: STREM CHEMICALS INC MS
Danger

93-5142-250G 93-5142-1000G
100208-578EA 128.8 USD
100208-578 100207-872
Antimony ≥99.5%, powder
Antimony
Formula: Sb
Boiling Pt: 1380 °C
Melting Pt: 630 °C
Density: 6.684
MDL Number: MFCD00134030
CAS Number: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR