Antimony ≥99.5%, powder
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-5134-2000G
93-5134-500G
93-5134-100G
100210-888EA
472.74
USD
100210-888
100205-640
100199-132
Antimony ≥99.5%, powder
Antimony
Formula:
Sb Boiling Pt: 1380 °C Melting Pt: 630 °C Density: 6.684 |
MDL Number:
MFCD00134030 CAS Number: 7440-36-0 EINECS: 231-146-5 UN: 2871 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...