Antimony ≥99.999%, powder
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-5133-5G
93-5133-25G
100207-102EA
179.75
USD
100207-102
100199-592
Antimony ≥99.999%, powder
Antimony
Formula:
Sb MW: 121.76 g/mol Boiling Pt: 1380 °C Melting Pt: 630 °C Density: 6.684 |
MDL Number:
MFCD00134030 CAS Number: 7440-36-0 EINECS: 231-146-5 UN: 2871 ADR: 6.1 III Merck Index: 12,00733 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...