Antimony(III) oxide ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Antimony trioxide, Diantimony trioxide, di-Antimony trioxide
93-5119-250G
93-5119-1000G
100205-636EA
86.34
USD
100205-636
100210-462
Antimony(III) oxide ≥99%
Antimony(III) oxide
Formula:
O₃Sb₂ MW: 291.52 g/mol Boiling Pt: 1425 (subl.) Melting Pt: 654 Density: 5.67 |
MDL Number:
MFCD00011214 CAS Number: 1309-64-4 EINECS: 215-175-0 UN: 1549 ADR: 6.1 III Merck Index: 13,00717 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...