Antimony(III) iodide ≥99.9% (Sb basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Antimony triiodide
93-5116-25G
93-5116-100G
93-5116-5G
100199-586EA
159.94
USD
100199-586
100210-460
100199-128
Antimony(III) iodide ≥99.9% (Sb basis)
Antimony(III) iodide
Formula:
I₃Sb MW: 502.47 Boiling Pt: 397 Melting Pt: 171 Density: 4.917 |
MDL Number:
MFCD00016322 CAS Number: 7790-44-5 EINECS: 232-205-8 UN: 1549 ADR: 6.1 III Merck Index: 12,00751 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...