Antimony (III) ethanolate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-5115-5G 93-5115-25G
100200-164EA 147.2 USD
100200-164 100210-696
Antimony (III) ethanolate ≥99%
Antimony (III) ethanolate

Alkoxide, Metal Alkoxides, Volatile Precursors for CVD

Formula: C₆H₁₅O₃Sb
MW: 256.94 g/mol
Boiling Pt: 95 °C (11 mmHg)
Density: 1.524
Flash Pt: 140 °F
MDL Number: MFCD00015123
CAS Number: 10433-06-4
EINECS: 233-914-5
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR