Antimony (III) butanolate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-5113-5G 93-5113-25G
100198-414EA 110.4 USD
100198-414 100203-504
Antimony (III) butanolate ≥99%
Antimony (III) butanolate

Alkoxide, Metal Alkoxides, Volatile Precursors for CVD

Formula: C₁₂H₂₇O₃Sb
Boiling Pt: 133…135 °C (4 mmHg)
Density: 1.23
MDL Number: MFCD00015238
CAS Number: 2155-74-0
EINECS: 218-456-6
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR