Antimony (V) oxide ≥99.9% (Sb basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Antimony pentoxide, Diantimony pentoxide

93-5106-10G 93-5106-50G
100205-634EA 84.92 USD
100205-634 100202-460
Antimony (V) oxide ≥99.9% (Sb basis)
Antimony (V) oxide
Formula: O₅Sb₂
MW: 323.52 g/mol
Melting Pt: 2000 °C
Density: 3.8 g/cm³ (20 °C)
MDL Number: MFCD00011216
CAS Number: 1314-60-9
EINECS: 215-237-7
UN: 1549
ADR: 6.1,III
Merck Index: 12,00739

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR