Tin ≥99.5%, powder
Supplier: STREM CHEMICALS INC MS
93-5089-500G
93-5089-100G
100198-410EA
225.05
USD
100198-410
100208-576
Tin ≥99.5%, powder
Tin
Formula:
Sn MW: 118.71 g/mol Boiling Pt: 2270 Melting Pt: 232 Density: 7.31 |
MDL Number:
MFCD00133862 CAS Number: 7440-31-5 EINECS: 231-141-8 UN: 3089 ADR: 4.1 III Merck Index: 12,09587 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...