Trimethyltin hydroxide ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-5085-10G
93-5085-2G
100207-098EA
532.18
USD
100207-098
100207-100
Trimethyltin hydroxide ≥98%
Trimethyltin hydroxide
Formula:
C₃H₁₀OSn MW: 180.82 g/mol Boiling Pt: 80 °C (subl.) Melting Pt: 118 °C |
MDL Number:
MFCD00013929 CAS Number: 56-24-6 UN: 3146 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...