Trimethyltin chloride ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

93-5084-5G 93-5084-25G
100197-884EA 62.28 USD
100197-884 100197-882
Trimethyltin chloride ≥98%
Trimethyltin chloride
Formula: C₃H₉ClSn
Boiling Pt: 154 °C
Melting Pt: 37 °C
Flash Pt: 207 °F
MDL Number: MFCD00000520
CAS Number: 1066-45-1
EINECS: 213-917-8
UN: 3146
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR