Tributyltin hydride ≥95%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Tri-n-butyltin hydride
93-5081-25G
93-5081-100G
100209-440EA
209.48
USD
100209-440
100205-630
Tributyltin hydride ≥95%
Tributyltin hydride
Formula:
C₁₂H₂₈Sn MW: 291.06 g/mol Boiling Pt: 78 °C (0.8 mmHg) Melting Pt: < 0 °C Density: 1.103 (20°) Flash Pt: 104 °F |
MDL Number:
MFCD00009416 CAS Number: 688-73-3 EINECS: 211-704-4 UN: 2929 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...