Tin(II) iodide ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tin(II) iodide, Tin diiodide

93-5077-5G 93-5077-25G
100196-720EA 60.86 USD
100196-720 100202-560
Tin(II) iodide ≥99%
Tin(II) iodide
Formula: I₂Sn
MW: 372.52 g/mol
Boiling Pt: 714 °C (1013 hPa)
Melting Pt: 320 °C
Density: 5.28 g/cm³ (25 °C)
MDL Number: MFCD00049545
CAS Number: 10294-70-9
EINECS: 233-667-3
UN: 3260
ADR: 8,III
Merck Index: 12,08942

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR