Tripropyltin chloride ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Tri-n-propyltin chloride, Chlorotripropylstannane
93-5063-25G
93-5063-5G
100201-124EA
901.6
USD
100201-124
100201-126
Tripropyltin chloride ≥98%
Tripropyltin chloride
Formula:
C₉H₂₁ClSn MW: 283.43 Boiling Pt: 234 Melting Pt: -23 Density: 1.2678 (28°) Flash Pt: 95 °C |
MDL Number:
MFCD00013610 CAS Number: 2279-76-7 EINECS: 218-910-3 UN: 2788 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...