Fentin fluoride ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-5061-25G
93-5061-5G
100210-446EA
244.86
USD
100210-446
100196-718
Fentin fluoride ≥97%
Fentin fluoride
Formula:
C₁₈H₁₅FSn MW: 369.03 g/mol Melting Pt: > 250 °C Density: 1.5 g/cm³ (20 °C) |
MDL Number:
MFCD00015356 CAS Number: 379-52-2 EINECS: 206-833-8 UN: 3146 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...