Fentin chloride ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Chlorotriphenylstannane, Triphenyltin chloride

93-5060-50G 93-5060-250G
100208-574EA 72.18 USD
100208-574 100203-858
Fentin chloride ≥95%
Fentin chloride
Formula: C₁₈H₁₅ClSn
MW: 385.48 g/mol
Boiling Pt: 249 °C (13.5 mmHg)
Melting Pt: 106 °C
Density: 1.49 g/cm³ (20 °C)
Flash Pt: >150 °C
MDL Number: MFCD00000519
CAS Number: 639-58-7
EINECS: 211-358-4
UN: 3146
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR