Tributyltin chloride ≥96%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Chlorotributylstannane, Tri-n-butyltin chloride, TBTC

93-5056-250G 93-5056-50G
100199-582EA 319.88 USD
100199-582 100205-624
Tributyltin chloride ≥96%
Tributyltin chloride
Formula: C₁₂H₂₇ClSn
MW: 325.51 g/mol
Boiling Pt: 172 °C (5 mmHg)
Melting Pt: -9
Density: 1.207 (20°)
Flash Pt: 210 °F
MDL Number: MFCD00000521
CAS Number: 1461-22-9
EINECS: 215-958-7
UN: 2788
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR