Tin(II) oxide ≥98% (99+%-Sn)

Supplier: STREM CHEMICALS INC MS

Synonyms: Stannous oxide, Tin monoxide, Tin(ii)oxide

93-5039-50G 93-5039-250G
100210-440EA 63.69 USD
100210-440 100205-886
Tin(II) oxide ≥98% (99+%-Sn)
Tin(II) oxide
Formula: OSn
MW: 134.71 g/mol
Boiling Pt: 1425
Melting Pt: 1040 Dec.
Density: 6.95
MDL Number: MFCD00011243
CAS Number: 21651-19-4
EINECS: 244-499-5
Merck Index: 12,08944

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR