Tin(II) oxalate

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tin (II) ethanedioate, Oxalic acid tin (II) salt

93-5038-50G 93-5038-250G
100202-176EA 39.63 USD
100202-176 100203-500
Tin(II) oxalate
Tin(II) oxalate
Formula: C₂O₄Sn
MW: 206.73 g/mol
Melting Pt: 280 °C (dec.)
Density: 3.56
Storage Temperature: Ambient
MDL Number: MFCD00040678
CAS Number: 814-94-8
EINECS: 212-414-0
UN: 3288
ADR: 6.1 III
Merck Index: 13,08865

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR