Tin(II) 2-ethylhexanoate ≥90% in 2-ethylhexanoic acid (~28% Sn)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-5037-1000G
93-5037-250G
100207-860EA
326.95
USD
100207-860
100210-438
Tin(II) 2-ethylhexanoate ≥90% in 2-ethylhexanoic acid (~28% Sn)
Tin(II) 2-ethylhexanoate
Formula:
C₁₆H₃₀O₄Sn Flash Pt: >287 °F |
MDL Number:
MFCD00002676 CAS Number: 301-10-0 EINECS: 206-108-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...