Tin(II) chloride dihydrate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Stannous chloride dihydrate
93-5031-500G
93-5031-100G
100197-876EA
162.77
USD
100197-876
100199-126
Tin(II) chloride dihydrate ≥98%
Tin(II) chloride dihydrate
Formula:
SnCl₂·2H₂O MW: 225.65 g/mol Boiling Pt: 623 °C (1013 hPa) Melting Pt: 246 °C Density: 2.71 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149863 CAS Number: 10025-69-1 EINECS: 231-868-0 UN: 3260 ADR: 8,III Merck Index: 13,08861 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...