Tin(IV) fluoride ≥98.5%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-5024-5G
93-5024-1G
100198-406EA
220.8
USD
100198-406
100196-706
Tin(IV) fluoride ≥98.5%
Tin(IV) fluoride
Formula:
SnF₄ MW: 194.7 g/mol Boiling Pt: 705 °C (1013 hPa) Melting Pt: 705 °C (subl.) Density: 4.78 g/cm³ (19 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00042541 CAS Number: 7783-62-2 EINECS: 232-016-0 UN: 1759 ADR: 8,II Merck Index: 12,08931 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...