Tin(IV) chloride pentahydrate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tin tetrachloride pentahydrate, Stannic chloride pentahydrate

93-5023-500G 93-5023-100G
100205-884EA 246.28 USD
100205-884 100210-432
Tin(IV) chloride pentahydrate ≥98%
Tin(IV) chloride pentahydrate
Formula: Cl₄Sn·5H₂O
MW: 350.6 g/mol
Melting Pt: ∼60 °C
Density: 2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149864
CAS Number: 10026-06-9
EINECS: 231-588-9
UN: 2440
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR