Tin(IV) chloride, anhydrous ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Stannic chloride, Tin tetrachloride

93-5022-1000G 93-5022-250G
100210-430EA 251.94 USD
100210-430 100205-618
Tin(IV) chloride, anhydrous ≥98%
Tin(IV) chloride
Formula: Cl₄Sn
MW: 260.52 g/mol
Boiling Pt: 114 °C (1013 hPa)
Melting Pt: -33 °C
Density: 2.22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011242
CAS Number: 7646-78-8
EINECS: 231-588-9
UN: 1827
ADR: 8,II
Merck Index: 12,08929

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR