Tin(IV) bromide ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tin tetrabromide, Stannic bromide

93-5021-100G 93-5021-25G
100207-484EA 294.4 USD
100207-484 100207-856
Tin(IV) bromide ≥99%
Tin(IV) bromide
Formula: Br₄Sn
MW: 438.33
Boiling Pt: 202
Melting Pt: 29.2
Density: 3.34
MDL Number: MFCD00011240
CAS Number: 7789-67-5
EINECS: 232-184-5
UN: 1760
ADR: 8 II
Merck Index: 12,08928

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR