Hexabutylditin ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Hexa-n-butylditin, Hexabutyldistannane, Bis(tributyltin)

93-5019-10G 93-5019-50G
100208-186EA 111.82 USD
100208-186 100205-882
Hexabutylditin ≥95%
Hexabutylditin
Formula: C₂₄H₅₄Sn₂
MW: 580.11 g/mol
Boiling Pt: 160
Density: 1.1480
Flash Pt: >230 °F
Storage Temperature: Ambient
MDL Number: MFCD00009417
CAS Number: 813-19-4
EINECS: 212-383-3
UN: 2788
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR