Diphenyltin sulfide ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-5018-5G 93-5018-1G
100198-404EA 184 USD
100198-404 100204-356
Diphenyltin sulfide ≥99%
Diphenyltin sulphide
Formula: C₁₂H₁₀SSn
MDL Number: MFCD00058856
CAS Number: 20332-10-9
EINECS: 243-739-6
UN: 3146
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR