Diphenyltin oxide ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Diphenyltin(IV) oxide, Diphenyloxostannane
93-5017-5G
93-5017-25G
100209-140EA
90.58
USD
100209-140
100199-282
Diphenyltin oxide ≥98%
Diphenyltin oxide
Formula:
C₁₂H₁₀OSn MW: 288.92 g/mol |
MDL Number:
MFCD00058851 CAS Number: 2273-51-0 EINECS: 218-883-8 UN: 3146 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...