Diphenyltin dichloride ≥96%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Dichlorodiphenylstannane

93-5016-10G 93-5016-50G
100196-704EA 90.58 USD
100196-704 100209-438
Diphenyltin dichloride ≥96%
Diphenyltin dichloride
Formula: C₁₂H₁₀Cl₂Sn
MW: 343.83 g/mol
Boiling Pt: 333…337 °C (dec.)
Melting Pt: 41…43 °C
MDL Number: MFCD00000516
CAS Number: 1135-99-5
EINECS: 214-496-3
UN: 3146
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR