Dimethyltin oxide ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-5014-5G
93-5014-25G
100199-578EA
164.18
USD
100199-578
100207-482
Dimethyltin oxide ≥97%
Dimethyltin oxide
Formula:
C₂H₆OSn Melting Pt: Dec. |
MDL Number:
MFCD00013838 CAS Number: 2273-45-2 EINECS: 218-881-7 UN: 3146 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...