Dibutyltin oxide ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Dibutyltin(IV) oxide, Dibutyloxostannane, Di-n-butyltin oxide, Di-n-butyltin(IV) oxide

93-5010-100G 93-5010-500G
100204-352EA 53.78 USD
100204-352 100199-576
Dibutyltin oxide ≥98%
Dibutyltin oxide
Formula: C₈H₁₈OSn
Melting Pt: Dec.
Density: 1.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001992
CAS Number: 818-08-6
EINECS: 212-449-1
UN: 3146
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR