Butyltin trichloride ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

93-5004-50G 93-5004-250G
100202-168EA 65.11 USD
100202-168 100209-136
Butyltin trichloride ≥95%
Butyltin trichloride
Formula: C₄H₉Cl₃Sn
Boiling Pt: 98 °C (10 mmHg)
Melting Pt: –63 °C
Density: 1.693
Flash Pt: 178 °F
MDL Number: MFCD00000515
CAS Number: 1118-46-3
EINECS: 214-263-6
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR