Bis(tributyltin) sulfide ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Bis(tri-n-butyltin)sulphide, Hexabutyldistannathiane

93-5002-25G 93-5002-5G 93-5002-100G
100197-246EA 852.06 USD
100197-246 101324-836 100208-568
Bis(tributyltin) sulfide ≥97%
Bis(tributyltin)sulphide
Formula: C₂₄H₅₄SSn₂
MW: 612.18 g/mol
Density: 1.197 g/cm³ (20 °C)
MDL Number: MFCD00048750
CAS Number: 4808-30-4
EINECS: 225-369-7
UN: 2788
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR