Indium ≥99.99%, powder
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-4940-25G
93-4940-5G
100202-706EA
532.18
USD
100202-706
100211-266
Indium ≥99.99%, powder
Indium
Formula:
In MW: 114.82 g/mol Boiling Pt: 2080 °C Melting Pt: 156.6 °C Density: 7.31 g/cm³ (20 °C) |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 UN: 3089 ADR: 4,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...