Indium(I) chloride, anhydrous ≥99.99% (In basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-4930-10G
93-4930-2G
100203-856EA
566.15
USD
100203-856
100199-574
Indium(I) chloride, anhydrous ≥99.99% (In basis), PURATREM
Indium(I) chloride
Formula:
InCl Boiling Pt: 608 °C Melting Pt: ∼225 °C Density: 4.19 g/cm³ (20 °C) |
MDL Number:
MFCD00148875 CAS Number: 13465-10-6 EINECS: 236-693-3 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...