Indium(III) bromide, anhydrous ≥99.999% (In basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Indium tribromide

93-4929-25G 93-4929-5G
100202-458EA 699.2 USD
100202-458 100205-614
Indium(III) bromide, anhydrous ≥99.999% (In basis), PURATREM
Indium(III) bromide
Formula: Br₃In
MW: 354.53 g/mol
Boiling Pt: 656
Melting Pt: 436
Density: 4.74
MDL Number: MFCD00011057
CAS Number: 13465-09-3
EINECS: 236-692-8
UN: 3260
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR