Diindium trisulfate hydrate ≥99.999% (In basis), PURATREM

Supplier: STREM CHEMICALS INC MS

93-4927-10G 93-4927-50G
100198-402EA 226.46 USD
100198-402 100209-134
Diindium trisulfate hydrate ≥99.999% (In basis), PURATREM
Diindium trisulphate hydrate
Formula: O₁₂In₂S₃·H₂O
Density: 3.44 g/cm³ (20 °C)
MDL Number: MFCD00149747
CAS Number: 207399-02-8
EINECS: 236-689-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR