Indium phosphide ≥99.999% (In basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Indium(III) phosphide, Indium phosphide
93-4926-1G
93-4926-5G
100197-242EA
332.62
USD
100197-242
100197-244
Indium phosphide ≥99.999% (In basis), PURATREM
Indium phosphide
Formula:
InP MW: 145.79 g/mol Melting Pt: 1070 °C Density: 4.487 g/cm³ (20 °C) |
MDL Number:
MFCD00016153 CAS Number: 22398-80-7 EINECS: 244-959-5 Merck Index: 13,04976 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...